Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Complete Liquid Sampling and Stirring Kit, Dynalon

Supplier: Dynalon

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

107035-5000
10053-760EA 709.02 USD InStock
10053-760
Complete Liquid Sampling and Stirring Kit, Dynalon
Samplers Fluid Samplers
These versatile modular design plastic liquid sampling and stirring kits will meet all of your needs

Kit components are manufactured of high density polyethylene (HDPE) and Polypropylene (PP). Their threaded connections allow quick and easy use. The Complete Liquid Sampling and Stirring Kit includes all ten kit components, items 10053-764 to 10053-752.

Complete Liquid Handling and Stirring Kit components are interchangeable. Component are also sold as separate items (See 10053-764, -766, -768, -750 and -752).

Whether sampling from drums, tanks, or rail cars, stirring product vats, taking water samples or dipping chemicals from a bath this complete kit configuration will help to meet all of your liquid handling, sampling and stirring requirements.

Bottle capacity: 100 mL, 250 mL, 500 mL, 1000 mL
Handle length: 1.5', 3', 6'
Weight: 7.2 lbs.

Certifications: FDA Compliant.

Ordering information: Kits and their components are available as one each.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty