Would you like to log in again?
Chocolate Agar with Bacitracin, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
E11 E11
10053-844PK 23.54 USD InStock
CA77497-276 10053-844
Chocolate Agar with Bacitracin, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Chocolate Agar with Bacitracin for Haemophilus isolation.
For selective isolation of Haemophilus spp. from respiratory specimens. Inhibits most staphylococci, streptococci, neisseria, and other normal flora.
Ordering information: Order by the pack of 10
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty