Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SAC-i Polarimeters, ATAGO®

Supplier: Atago
5951
10054-096EA 31672.75 USD InStock
10054-096
SAC-i Polarimeters, ATAGO®
Polarimeters
ATAGO's Polarimeters measure the optical rotation of optically active substances such as saccharide, ascorbic acid, sodium glutamate and others.

  • Measurement stability in 13 seconds (4 seconds in the high-speed and consecutive measurement mode)
  • Resolution of up to 3 decimal places
  • Repeatability of ±0.003° Angle of Rotation and ±0.009°Z
  • Up to 999 automatic consecutive measurements followed by average value display
  • Sleep and timer feature to take measurement later at specified time
  • Improved usability with the use of touch screen technology
  • Connectivity to RX Digital Refractometers for automatic purity measurement

Angle of rotation measurement: -89.999 - +90.000° or -360 - +360°
Angle of rotation accuracy: ±0.005° (-45 - +45°)
Angle of rotation resolution: 0.001°

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty