Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RT2 Basic Hotplate Stirrer, Thermo Scientific

Supplier: Thermo Scientific
88880004
10054-156EA 1024.39 USD InStock
10054-156
RT2 Basic Hotplate Stirrer, Thermo Scientific
Stirrers Magnetic Hotplate Stirrers
The reliable accuracy of these sleek, low-profile, analog hotplate stirrers can be relied upon for routine applications.

  • In event of viscosity change, the feedback function maintains regular speed
  • Hot top warning indicator
  • Automatic over temperature safety cut-off at 450 °C

A consistent stirring speed up to 2000 rpm is achieved with the feedback control design of the RT2 Basic series. It provides constant stirring speed even when the viscosity of the media changes during operation. The RT2 basic features exceptionally strong magnetic coupling power and a smooth-start stirring mechanism. Virtually no decoupling of magnetic stir bars occurs, even with viscous media or at high speeds. The unit boasts quick heat-up time due to it's ample heating capacity (600 W), as well as a superb heat transfer rate enabled by the tightly integrated structure of the heater and the ceramic-coated aluminum alloy top plate. The top provides exceptional temperature uniformity, while the thin white ceramic coating provides excellent corrosion resistance. This combination allows for optimal monitoring of color changes in samples or media during operation. Safety features include hot top warning (even when unit is shut off and still plugged in), automatic over-temperature shut-off, detachable power cord, and optional transparent shield. Unit includes two threaded holes for support rods to hold various devices, such as temperature sensors, thermometers, laboratory glassware,and more.

Max load: 25 kg (55.1 lbs.)

Certifications: CE marked. cCSAus approved.

Ordering information: 2-year warranty included.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty