Would you like to log in again?
Parter Medical Products Petri Dish, Hardy Diagnostics
Supplier: Hardy Diagnostics
3536 3561 3536 3304
10054-172CS 180.62 USD InStock
10054-172 CA10790-408 10790-408 10054-174
Parter Medical Products Petri Dish, Hardy Diagnostics
Culture Dishes Petri Dishes
Optically clear polystyrene - for distortion free viewing.
Hardy offers a full selection of petri plates:
Mono plate
Space Saver Plate
Bi-Plate
Tri-Plate
Quad Plate
Membrane Filter Methods
Contacts Plate
Heavy Duty - reduces breakage during shipping.
Stacking rings - guards against slippage when moving stacked plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty