Would you like to log in again?
TouchNTuff® 73-300 Neoprene Gloves, Non-Sterile, Ansell
Supplier: Ansell
TouchNTuff®
73300080 73300055 73300060 73300070 73300075 73300065 73300090
10054-568CS 1001.73 USD InStock
10054-568 10054-578 10054-576 10054-574 10054-572 10054-582 10054-570
TouchNTuff® 73-300 Neoprene Gloves, Non-Sterile, Ansell
Gloves
The TouchNTuff® 73-300 neoprene gloves deliver superior sensitivity and comfort in nonsterile, clean environments.
  • Hand-specific
  • Great splash resistance against a broad range of chemicals
  • Ergonomically designed to enhance fit and comfort
  • Reduced hand fatigue and stress
These sensitive neoprene gloves are a new feel in double-donning and dry applications.
TouchNTuff® single-use gloves provide superior tactility and high resilience in wet and dry industrial, lab, and controlled environments. Gloves are packed in a controlled environment, in a cleanroom-compatible packaging material, which is printed with IPA-resistant ink.
Customers need to make their own determination based on cleanliness standards. Ansell expects this glove should be suitable for ISO Class 5 or higher cleanroom environments.
The exceptionally thin polychloroprene formulation reduces the risk of Type I or IV allergies to the wearer caused by natural rubber latex or other synthetic gloves. The sensitive neoprene gloves offer users splash resistance against a broad range of chemicals. Ansell’s SureFit Technology™ eliminates 'cuff roll down' by maintaining gloves securely around the arm, providing wearer with more protection during routine lab work and manufacturing by ensuring the efficacy of the gloves as a barrier protection. Gloves are ergonomically designed and hand-specific to enhance fit and comfort, and to reduce hand fatigue and stress.
Certifications: Audit Standards Manufacturing Facility Registered to ISO 9001, ISO 13485 and ISO 14001. Quality Control Compliant with ASTM D 3577, Type II, Cat. III glove.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty