Would you like to log in again?
Dry Block Heaters, IKA® Works
Supplier: IKA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0004025401 0004025201 0004025101 0004025301
10054-600EA 2869.4 USD InStock
10054-600 10054-598 10054-596 10054-594
Dry Block Heaters, IKA® Works
Block Heaters Digital Block Heaters
Digital dry block heaters with precisely designed blocks ensure highly accurate and reproducible results, guaranteeing short heating times
IKA® dry block heaters are highly versatile and each model can accept separate interchangeable modular heating blocks. They can accommodate various sample enclosures such as centrifuge tubes, test tubes, vials, microtitre plates, and PCR tubes or strips. Each block has a thermometer well for measuring the block temperature. Anodized aluminum blocks provide superior temperature stability and heat transfer. A PT 1000.60 temperature sensor included in the delivery.
The bright, easy to read LED display facilitates very easy setting of the required temperature and accurate monitoring of the actual temperature. As a result, units offer excellent temperature uniformity and stability.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty