Would you like to log in again?
TouchNTuff® 73-500 Neoprene Gloves, Sterile, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TouchNTuff®
73500070 73500090 73500085 73500060 73500065 73500080 73500075 73500055
10055-027CS 553.18 USD InStock
10055-027 10054-589 10054-587 10055-023 10055-025 10054-585 10054-595 10054-593
TouchNTuff® 73-500 Neoprene Gloves, Sterile, Ansell
Gloves
The TouchNTuff® 73-500 neoprene gloves deliver superior sensitivity and comfort in sterile, aseptic environments.
  • These sensitive neoprene gloves are a new feel in double-donning and dry applications
  • TouchNTuff® single-use gloves provide superior tactility
  • High resilience in wet and dry industrial, lab, and controlled environments
Gloves are packed in a controlled environment, in a cleanroom-compatible packaging material, which is printed with IPA-resistant ink.
Customers need to make their own determination based on cleanliness standards. Ansell expects this glove should be suitable for ISO Class 5 or higher cleanroom environments.
The exceptionally thin polychloroprene formulation reduces the risk of Type I or IV allergies to the wearer caused by natural rubber latex or other synthetic gloves. The sensitive neoprene gloves offer users splash resistance against a broad range of chemicals. SureFit Technology™ eliminates 'cuff roll down' by maintaining gloves securely around the arm, ensuring the efficacy of the gloves as a barrier protection. Gloves are ergonomically-designed and hand specific to enhance fit and comfort, and to reduce hand fatigue and stress.
Certifications: Audit Standards Manufacturing Facility Registered to ISO 9001, ISO 13485 and ISO 14001. Quality Control Compliant with ASTM D 3577, Type II, Cat. III glove.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty