Would you like to log in again?
Thermo Scientific Super Clean Cartridge Filter Baseplates, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
60180-815 60180-818 60180-814 60180-817
10055-290EA 1002.94 USD InStock
10055-290 10055-302 10055-288 10055-296
Thermo Scientific Super Clean Cartridge Filter Baseplates, Thermo Scientific
Filters In-Line Filters
A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change.
Baseplates require one-time installation only.
Delivery information: Super Clean Cartridge Filters are not included.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty