Would you like to log in again?
Reacti-Vap Evaporators, Thermo Scientific
Supplier: Thermo Scientific
TS-18826 TS-18825
10055-298EA 1372.8 USD InStock
10055-298 10055-300
Reacti-Vap Evaporators, Thermo Scientific
Evaporators Centrifugal Evaporators
Manifolds for easy sample evaporation, with integrated pressure-relief valve to protect against excessive gas flow and dangerous pressure build up
Easy set up: attach to corresponding Reacti-Therm Module, attach tubing from gas supply, and lower into position over samples and start gas flow.
Ordering information: For a list of compatible aluminum alloy blocks, please see Reacti-Block Aluminum Blocks, Thermo Scientific.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty