Would you like to log in again?
Ultra-Precision NMR Sample Tubes, 5 mm, NORELL
Supplier: Norell
S-5-300-7 S-5-500-7 S-5-800-8 S-5-900-7 S-5-900-8 S-5-500-8 S-5-400-8 S-5-600-7 S-5-400-7 S-5-300-8 S-5-600-8 S-5-600-VT-8
100554-200PK 391.43 USD InStock
100554-200 100554-212 100554-202 100554-214 100554-204 100554-216 100554-206 100554-208 100554-194 100554-196 100554-198 100554-210
Ultra-Precision NMR Sample Tubes, 5 mm, NORELL
Tubes NMR Tubes
These NMR tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex® 7740 (Corning), Duran® (Schott Glass), or Kimax® KG-33 (Kimble) glass.
Key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient.
This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or under any other application where large temperature variations are required in the experiment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty