Would you like to log in again?
PYREX TUBE, 3MM, 900MHZ, 7IN
Supplier: NORELL INC MS
Select Series NMR Sample Tube^ Designed for high-resolution NMR. Pyrex, 3MM, 900 MHz, Round Bottom 7/ Caps Included. OD: 2.99 +/-0.004mm ID: 2.41 +/-0.006mm Concentricity: 0.0020mm CAMBER: +/-0.0030mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PYREX TUBE, 3MM, 900MHZ, 7IN 100554-218 S-3-900-7 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty