Would you like to log in again?
TracePLOT TG-BOND Alumina GC Columns Na₂SO₄ Deactivation, Thermo Scientific
Supplier: Thermo Scientific
TracePLOT™
26001-6080 26001-6020 26001-6110 26001-6050
10056-006EA 1398.69 USD InStock
10056-006 10055-932 10056-004 10056-008
TracePLOT TG-BOND Alumina GC Columns Na₂SO₄ Deactivation, Thermo Scientific
Chromatography Columns
Optimized for linear and quantitative analysis of polar unsaturated hydrocarbons.
  • Strong bonding to prevent particle generation
  • Columns to which water has adsorbed may be regenerated to restore full efficiency and selectivity
Each column has been tested to ensure proper film thickness (1,3-butadiene), selectivity (propadiene and methyl acetylene), resolution (trans-2-butene and 1-butene) and coating efficiency (1,3-butadiene).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty