Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermo Scientific Click-On Inline Filters, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Click-On™
60180-802 60180-808 60180-804 60180-801 60180-805 60180-806 60180-803 60180-843
10055-998EA 246.63 USD InStock
10055-998 10056-042 10056-074 10056-294 10056-002 10056-000 10055-164 10056-076
Thermo Scientific Click-On Inline Filters, Thermo Scientific
Filters In-Line Filters
This in-line filter removes contamination from gas with easy-to-use filters. Reuseable end-fittings containing needle valves remain installed during filter changes, preventing contamination and reducing the cost of clean gas.

  • Pure gas output 99.9999% or 6.0 grade
  • No carrier gas line contamination during filter change
  • Easy and fast replacement without the need for tools

Max. pressure: 11 bar

Certifications: TUEV approved.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty