Would you like to log in again?
TraceGOLD™ GC Columns with SafeGuard, Thermo Scientific
Supplier: Thermo Scientific
26096-1421 26088-1421 26098-1435 26099-2985 26099-1425 26096-2235 26096-1425 26098-5785 26098-1425 26098-1305 26096-1301 26098-0475 26088-1435
10056-688EA 1201.7 USD InStock
10056-688 10056-676 10056-388 10056-604 10056-758 10056-746 10056-800 10056-426 10056-680 10056-692 10055-730 10056-606 10056-616
TraceGOLD™ GC Columns with SafeGuard, Thermo Scientific
Chromatography Columns
Extend your column lifetime without compromising performance.
SafeGuard is an integrated guard on the GC column.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty