Would you like to log in again?
TraceGOLD™ TG-35MS GC Columns
Supplier: Thermo Scientific
TraceGOLD™
26094-3360 26094-2240 26094-1420 26094-2980 26094-1430 26094-1300 26094-2230 26094-5780 26094-2130
10056-556EA 1074.48 USD InStock
10056-556 10056-566 10056-554 10056-564 75814-688 10056-580 10056-584 10056-562 10056-550
TraceGOLD™ TG-35MS GC Columns
Chromatography Columns
These GC columns have a higher phenyl content for useful elution order and retention time changes.
  • Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
  • Equivalent to USP G42 phase
Ideal for organochlorine pesticides and herbicides, pharmaceuticals, sterols, phthalate esters etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty