Would you like to log in again?
TraceGOLD™ TG-624SiLMS GC Columns
Supplier: Thermo Scientific
TraceGOLD™
26059-3410 26059-4080 26059-3320 26059-4950 26059-3330 26059-3960 26059-3390
10056-820EA 1575.79 USD InStock
10056-820 10056-644 10056-648 10057-138 10056-646 10056-816 10056-818
TraceGOLD™ TG-624SiLMS GC Columns
Chromatography Columns
Analyze residual solvents, volatile organic compounds, alcohols, and oxygenates using Thermo Scientific™ TraceGOLD™ TG-624SilMS GC Columns. Both phases offer the low to mid polarity phase, 6% cyanopropylphenyl methypolysilozane.
TraceGOLD TG-624SilMS GC Columns provide high thermal stability with maximum temperatures up to 320 °C, which is ideal for volatile organics analysis. They are highly inert, providing excellent peak shape for a wide range of compounds.
Applications includes Residual solvents, volatile organic compounds, alcohols, oxygenates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty