Would you like to log in again?
TracePLOT™ Particle Traps for GC Instruments, Thermo Scientific
Supplier: Thermo Scientific
60180-860 60180-861
10056-884EA 264.31 USD InStock
10056-884 10056-296
TracePLOT™ Particle Traps for GC Instruments, Thermo Scientific
Chromatography Columns
Safeguard GC instruments from damage caused by dislodged particles entering the detector with Thermo Scientific TracePLOT Particle Traps for GC Instruments.
Ordering information: Traps include two connectors, 2.5 m column.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty