Would you like to log in again?
TRACE TR-1701 GC Columns, Thermo Scientific
Supplier: Thermo Scientific
260Q143P 260Q298P 260Q142P
10056-950EA 1132.37 USD InStock
10056-950 10056-948 10056-946
TRACE TR-1701 GC Columns, Thermo Scientific
Chromatography Columns
Mid-polarity column with alternative selectivity.
Low bleed even at a high operating temperature. Excellent starting point for method development. Suitable for a wide variety of applications. Recommended for pesticides, PCB's, PAH's, Organic Acids, Drugs, Steroids, and EPA 608, 8081.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty