Would you like to log in again?
TRACE TR-5HT GC Columns, Thermo Scientific
Supplier: Thermo Scientific
260H035P 260H142P 260H047P 260H030P
10057-026EA 1031.17 USD InStock
10057-026 10057-028 10057-030 10057-032
TRACE TR-5HT GC Columns, Thermo Scientific
Chromatography Columns
Feature upper temperature limits as high as 400 ˚C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty