Would you like to log in again?
TMSI Silylation Reagents
Supplier: Thermo Scientific
TS-88623 TS-88625
10057-398EA 407.08 USD InStock
10057-398 10066-410
TMSI Silylation Reagents
Chromatography Sorbents GC Sorbents
Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI silylation reagent.
  • Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
  • Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
  • Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
  • Excellent for C1 through C5 fatty acids in serum and urine
This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1 to C5 fatty acids in serum and urine.
Standard concentration: 98 to 100%
Hazards: Flammable, irritant
Standard components: 1-(Trimethylsilyl)-1 H-imidazole
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty