Would you like to log in again?
SilTite® μ-Unions, Gas Chromatography Connections
Supplier: Thermo Scientific
SilTite®
290SM301 290SM303 290SM403 290SM322 290SM405 290SM305 290SM401 290SM321 290SM302 290SM404 290SM304
10057-278EA 212.69 USD InStock
10057-278 10057-454 10057-464 10057-456 10057-448 10057-282 10057-450 10057-284 10057-460 10057-452 10057-276
SilTite® μ-Unions, Gas Chromatography Connections
Fittings and Connectors
The SilTite μ-Union is a connector for GC capillary columns, giving zero dead volume. The product has low thermal mass - it is only 9 mm in length and has a mass <0.5 g. It is available in kits to connect columns from 0.1 to 0.53 mm ID.
  • Zero dead volume - giving optimized peak shapes
  • FingerTite technology - easy to install and leak-free
  • Highly inert and robust
Ordering information: Kit contains 5 × ferrules, 2 × union fittings, Installation jigs, Installation instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty