Would you like to log in again?
MONITOR BL GR 0.8UM ST 47MM PK50
Supplier: GVS FILTER TECHNOLOGY
Microbiological Monitor 100ml^ Sterile Microbiological Monitor with Black Cellulose Nitrate Membrane and White Grid, Pore Size: 0.8um, Diameter: 4.7cm, Specifically for membrane filter methods for aqueous samples of up to 100mL.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MONITOR BL GR 0.8UM ST 47MM PK50 10057-612 10497503 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty