Would you like to log in again?
CENTREX NYLON 0.45UM 5ML ST PK50
Supplier: GVS FILTER TECHNOLOGY
Filter^ Centrex* centrifuge filters, Nylonmembrane, 0.45um,White, 5 ml, sterile, pk50, Reduced contamination risk with radioactive biologically hazardous material, Ideal for automated systems and high-speed batch filtration with robots, More than six samples can be processed at once
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CENTREX NYLON 0.45UM 5ML ST PK50 10057-754 10467021 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty