Would you like to log in again?
Axygen® Automation Tips for Hamilton® Microlab® Star Line and Nimbus®, Corning
Supplier: Corning
Axygen®
HT-50-C-HTR-5 HTF-50-C-HTR-S-5 HT-300-CBK-HTR HT-50-CBK-HTR HTF-1000-CBK-HTR-S HT-10-C-HTR-S-5 HT-10-CBK-HTR-5 HT-10-CBK-HTR HT-50-CBK-HTR-5 HT-10-C-STK-S HT-1000-C-HTR-S-5 HT-300-C-STK HTF-10-CBK-HTR-S HT-50-C-HTR-5 HT-50-C-HTR-S-5 HT-10-C-STK HT-50-CBK-HTR-S-5 HT-50-CBK-STK-S HT-1000-C-HTR-5 HT-300-C-HTR-5 HT-300-CBK-HTR-S HT-10-C-HTR-S-5 HTF-50-C-HTR-S-5 HT-1000-C-HTR-S-5 HT-10-CBK-STK-S HT-300-CBK-STK HT-1000-C-HTR-5 HT-50-C-STK-S HTF-1000-CBK-HTR-S HT-300-C-STK HTF-10-C-HTR-S-5 HTF-50-CBK-HTR-S-5 HTF-10-C-HTR-S-5 HT-300-CBK-STK-S HT-50-CBK-STK-S HT-50-CBK-STK HT-10-C-HTR-5 HT-1000-CBK-HTRS5 HTF-300-C-HTR-S-5 HT-300-C-HTR-5 HT-10-CBK-HTR-S HTF-300-CBK-HTR-S HT-300-CBK-STK-S HT-50-CBK-HTR-S HT-10-CBK-HTR-S-5 HT-300-CBK-HTR-S-5 HT-10-C-HTR-5 HT-10-C-STK HTF-300-CBK-HTRS5 HT-10-C-STK-S HT-300-C-HTR-S-5 HT-1000-CBK-HTR-5 HTF-1000-C-HTR-S-5 HT-300-CBK-HTR-5 HT-300-C-HTR-S-5 HTF-50-CBK-HTR-S HT-50-C-STK HT-10-CBK-STK-S HT-50-C-STK-S HT-50-C-STK HTF-1000-C-HTR-S-5 HTF-300-C-HTR-S-5 HTF-1000-CBKHTRS5 HTF-10-CBK-HTR-S-5
89230-166CS 377.53 USD InStock
89230-166 10057-868 10127-414 10057-872 10057-874 10127-416 CA10127-505 10057-876 10060-160 10057-877 10060-162 CA10127-509 CA10127-507 89230-162 10057-870 89230-160 89230-132 10127-510 CA10159-078 10127-504 CA10127-511 10127-506 CA10159-074 10127-508 CA10159-076 10693-086 89230-170 CA10159-072 89230-130 10693-088 10693-090 89230-142 89230-148 CA10127-403 10159-076 CA10127-407 10159-078 CA10127-405 10159-072 10159-074 CA10127-409 89230-140 CA10159-080 89230-158 10057-858 CA10746-462 10127-412 CA10746-460 CA10127-413 10057-860 10127-402 CA10127-411 10057-862 10127-404 CA10127-417 10127-406 10057-864 CA10746-458 CA10127-415 10127-408 10057-866 89230-152 10159-080 89230-150
Axygen® Automation Tips for Hamilton® Microlab® Star Line and Nimbus®, Corning
Pipette Tips
Axygen brand automation tips are compatible with Hamilton® Microlab® Star and Nimbus® robotic systems.
  • Conductive black or clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
Tips are available in 4 sizes (10, 50, 300, and 1000 µl) and are packaged in either traditional hanging tip racks ('frame' style) or deck-seating nested tip racks. As with all Axygen robotic tips, they are manufactured to stringent specifications under strict process control and assembled via automation to ensure consistent performance and quality.
Ordering information: Tips are also available in five tray blister packs with barcoded trays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty