Would you like to log in again?
Ferrules for Thermo Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: RESTEK CORP
20282 20283
10057-902EA 121.56 USD InStock
10057-902 10852-552
Ferrules for Thermo Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
For use with Thermo Scientific GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.25 mm or 0.32 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty