Would you like to log in again?
Front Ferrule, Swagelok®, Restek
Supplier: Restek
Swagelok®
23155 23103 23104 23154 23105 23153
10058-872PK 82.61 USD InStock
10058-872 10855-472 10058-874 10057-920 10057-922 10058-876
Front Ferrule, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
No coating required, but ferrules can be treated upon request (custom parts).
Ordering information: Note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty