Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferrules for Shimadzu 17A, 2010, and 2014 GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek

Supplier: RESTEK CORP
24828
10057-968PK 143.99 USD InStock
10057-968
Ferrules for Shimadzu 17A, 2010, and 2014 GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Fits columns with internal diameters of 0.25 mm or 0.32 mm

  • Graphite two-piece construction
  • Available in 0.4, 0.5, and 0.8 mm sizes

For use with Shimadzu 17A, 2010, and 2014 GCs.

Packaging: Packaged on mandrel for easy handling.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty