Would you like to log in again?
Polar ISO Column Test Mix, Restek
Supplier: Restek
35103
10058-062EA 46.44 USD InStock
10058-062
Polar ISO Column Test Mix, Restek
Standards Organic Standards
Polar ISO column test mix includes aniline, methyl dodecanoate, 2-chlorophenol, n-nonadecane, 2-dodecanol, 2-nonanone, n-heptadecane, 1-octanol.
  • 250 μg/mL each in 1,2-dichloroethane, 1 mL/ampoule
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty