Would you like to log in again?
Rxi®-1ms Capillary Columns, Restek
Supplier: Restek
Rxi®
13340 13342 13353 43801 13399 13350 13370 13321 13323 13320 13397 13311 13336 13367 13338 13335 13337 43802 13339 13301 13357 13320-600 13373 13341 13354 13356 13355 13324 43800 13327 13326 13396 13398 13302 13352 13397-600
89047-956EA 475.79 USD InStock
89047-956 89047-978 89047-976 89047-954 89047-998 89047-952 89047-974 89047-996 89047-972 89047-994 89047-950 89047-970 89047-992 89047-990 10852-088 10852-022 89048-006 89048-008 89048-002 89048-004 89048-000 89047-968 89047-988 89047-966 89047-964 89047-986 89047-962 89047-982 89047-960 89047-980 10058-386 10850-714 89048-010 10059-554 10059-552 89047-958
Rxi®-1ms Capillary Columns, Restek
Chromatography Columns
Inert, low-bleed columns for reliable column-to-column results from low-level GC/MS analyses provide reproducible retention and selectivity.
  • Low bleed
  • Reliable column-to-column results
  • General-purpose columns
  • Inert properties
Designed for drugs, essential oils, hydrocarbons, pesticides, PCB congeners, sulfur compounds, amines, oxygenates, gasoline range organics (GRO), and other general-purpose analyses. Equivalent to USP G2 Phase. Phase composition: Crossbond® 100% dimethyl polysiloxane. Temperature range: −60 to 330/350 °C (−76 to 626/662 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty