Would you like to log in again?
Rxi®-17Sil MS Columns (Fused Silica), Restek
Supplier: Restek
14124 14102 14123 14111 14126 43820 14121 14120 43821
10059-560EA 444.13 USD InStock
10059-560 10058-486 89205-682 10058-488 10058-490 10058-492 10058-484 10849-704 10059-562
Rxi®-17Sil MS Columns (Fused Silica), Restek
Chromatography Columns
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
  • Excellent inertness and selectivity for active environmental compounds, such as PAHs
  • Low bleed for use with sensitive detectors, such as MS
  • 340/360 °C upper temperature limits
  • Equivalent to USP phase G3
  • Mid-polarity Crossbond® phase
In addition to outstanding performance, this combination of column characteristics assures Rxi® columns will have long lifetimes. Restek has also developed a stable, reproducible manufacturing process, so that Rxi® columns will meet your needs every time you install a new column.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty