Would you like to log in again?
Restek Enhanced Capillary Nut for Shimadzu 17A, 2010, and 2014 GCs, Restek
Supplier: Restek
20375
10058-558PK 84.9 USD InStock
10058-558
Restek Enhanced Capillary Nut for Shimadzu 17A, 2010, and 2014 GCs, Restek
Fittings and Connectors
Restek's enhanced design for the capillary nut eliminates the slot, increasing lifetime and durability.
  • All parts meet or exceed original manufacturer's performance
For use with Shimadzu 17A, 2010, and 2014 GCs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty