Would you like to log in again?
PTFE Ferrules, Restek
Supplier: Restek
21125 21124 21123 21126 21127 21128 21122
10852-616PK 34.5 USD InStock
10852-616 10058-610 10852-614 10058-604 10852-622 10058-606 10058-608
PTFE Ferrules, Restek
Fittings and Connectors
PTFE ferrules are built to be extremely durable, with an inert nature and temperature resistance up to 250°C.
  • 100% PTFE fabrication
  • Completely inert
  • One-piece design requires no back ferrule
  • Upper temperature limit of 250°C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty