Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tedlar® Sampling Bags, Restek
Supplier: Restek
Tedlar®
22056 22054 22057 22053 22058 22049 22055 22051 22052 22050
10863-102PK 447.51 USD InStock
10863-102 10862-524 10863-026 10803-272 10862-764 10862-610 10058-714 10803-190 10862-806 10862-804
Tedlar® Sampling Bags, Restek
Bags Gas Bags
Sampling bags are a low-cost whole-air sampling device for high-level VOCs and permanent gases.
  • Find the bags you need - we offer sizes from 0.5 to 100 L
  • Unique all-in-one septum and valve fitting make these lightweight and easy to use
  • Polypropylene valve conveniently connects to ³/₁₆" Int.Ø PTFE tubing
  • Continuous sampling temperature up to 107 °C (225 °F); short term (1 to 2 hours) temperature up to 176 °C (350 °F)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for your application. All Restek's bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty