Would you like to log in again?
Zero Dead Volume Tee, Valco®, Restek
Supplier: Restek
Valco®
22535 22534
10058-784EA 155.53 USD InStock
10058-784 10855-612
Zero Dead Volume Tee, Valco®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert
Use these tee connectors for making three-way connections with tubing. Available with connection sizes of 1/16" or 1/8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty