Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Gas Sample Loops (Sulfinert®-Treated), Restek
Supplier: Restek
22846 22847 22848 22843 22842 22849 22841 22850 22845 22844
10058-820EA 265.16 USD InStock
10058-820 10058-810 10058-812 10058-814 10058-804 10058-816 10058-806 10058-818 10803-302 10058-808
Gas Sample Loops (Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Sulfinert® treatment eliminates active sites in the valve or loop for better recovery of active compounds.
  • Ideal for samples containing low concentrations of sulfur or other active compounds
  • Sample loop sizes from 5 µL to 5 mL
These gas sample loops have fittings sized at 1/16" with an internal diameter of 1 mm, for use with “W Type” valves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty