Would you like to log in again?
Metric 9-Piece, Ball-Point Hex Key Set, Restek
Supplier: Restek
22999
10058-834EA 56.97 USD InStock
10058-834
Metric 9-Piece, Ball-Point Hex Key Set, Restek
Wrenches
Ball-point hex key style includes nine metric hex keys (Allen wrenches): 1.5, 2, 2.5, 3, 4, 5, 6, 8, and 10 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty