Would you like to log in again?
Valco® Adapter Ferrules, ¹/₃₂", Valcon Polyimide, Restek
Supplier: Restek
Valco®
20140 20141 20137
10803-208EA 103.49 USD InStock
10803-208 10852-926 10059-058
Valco® Adapter Ferrules, ¹/₃₂", Valcon Polyimide, Restek
Fittings and Connectors
Fused silica adaptor ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C.
  • Durable
  • Fabricated from Valcon polyimide
The determining factor for selecting adapter ferrule size is the fused silica tubing OD.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty