Would you like to log in again?
Fixing Nut for Capillary Column for Split/Splitless Injector on Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL, Restek
Supplier: RESTEK CORP
24973
10059-334PK 56.87 USD InStock
10059-334
Fixing Nut for Capillary Column for Split/Splitless Injector on Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL, Restek
Fittings and Connectors
For use with split/splitless injectors on Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL.
  • High-quality stainless steel construction
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty