Would you like to log in again?
Waste Systems, Canister Kits, S.C.A.T.
Supplier: S.C.A.T.
107307 107310 107328
10060-708EA 430.38 USD InStock
10060-708 10060-710 10060-712
Waste Systems, Canister Kits, S.C.A.T.
Safety Cans
Complete waste systems including canister, SafetyWasteCap and exhaust filter.
  • Delivery is made complete, system is immediately ready for use
  • Only one article number for the comprehensive safety system
  • No time consuming compilation of a configuration
  • Price advantage compared to ordering piece by piece
All kits provide small exhaust filter, 2× capillary connector with 2.3/3.2 mm Ext.Ø, and 1× tube connector 6.4 to 9.0 mm interior diameter. The 5 and 10 L canister kits come with a SafetyWasteCap GL45 and the Space-Saving Canister Kit comes with SafetyWasteCap S50.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty