Would you like to log in again?
Autosampler Vials, Headspace, DWK Life Sciences
Supplier: DWK Life Sciences
90S332375BMB 90S332346B 90S332375BMB
10061-490CS 504.26 USD InStock
10061-490 10061-484 10061-488
Autosampler Vials, Headspace, DWK Life Sciences
Vials
Round bottom vials distribute the internal pressure created at high temperatures across the glass surface and are more easily handled by robotic arms that lift the vial from the tray.
  • Designed to fit most headspace autosamplers
  • Flat finish on top and bottom to maximise heating efficiency
The beveled edge 20 mm crimp seal finish features a sturdy rim that presses into the septum for a more effective seal.
Certifications: Meets or exceeds OEM specifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty