Would you like to log in again?
RECORDER TEMP/RH 12 INCHES TRULINE
Supplier: CARON PRODUCTS & SERVICE
Temperature/Humidity Recorder^ Side-Mounted Temperature/Humidity Honeywell Truline Recorder, Thermal Printer, 12inch , 7 Day, 24 Hour, for 7000 series units.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RECORDER TEMP/RH 12 INCHES TRULINE 10062-728 RCDR323 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty