Would you like to log in again?
HyFlex® 11-518 Ultralight-Duty Cut Protection Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
111710 111712 111711 235134 235135 111709 235131 111708 111707 235132 235133 235130
10062-922CS 2356.44 USD InStock
CA10062-920 10062-922 CA10062-922 10062-924 CA10062-924 10062-926 10062-916 CA10062-926 10062-918 CA10062-916 CA10062-918 10062-920
HyFlex® 11-518 Ultralight-Duty Cut Protection Gloves, Ansell
Gloves
A unique combination of engineered materials makes these the first gloves with 18-gauge comfort and ANSI Cut Level 2 (EN388 Cut Level 3) performance.
  • Enhanced flexibility
  • Superior tactile sensitivity
  • Breathable and lightweight material
  • Excellent resistance to abrasions
The ultralight-duty 18-gauge seamless liner delivers enhanced flexibility (range of motion) and tactility (fingertip sensitivity).
Ansell’s D3 Pinky Knitting tailors the fit at the base of the pinky finger to better match natural hand contours. Sizes 6-11 ensure optimum fit and comfort for every worker. The 11-518 cut-resistant glove belongs to next-generation of HyFlex® styles that bring legendary HyFlex® performance to an ultralight fabric with a barehand-like sensation of tactility and dexterity.
Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
Use in the automotive and transportation; logistics and warehousing; machinery and equipment; and metal fabrication industries. Uses include final assembly; handling and assembly of small or ultra-fine sharp parts; injection molding parts assembly; machining special metal blends; packaging areas with risk of cut; light and dry stamping; and wire assembly and fastening.
Certifications: ANSI Level 3 abrasion protection; ANSI Level 2 cut protection.
Packaging: Gloves are packaged 12 pairs per polybag; 144 pairs per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty