Would you like to log in again?
Femtogram Sensitive Chemiluminescence Detection, G-Biosciences
Supplier: G-Biosciences
786-10APT 786-056 786-10 786-10T 786-056 786-10 786-10APT 786-081 786-10AP 786-003 786-10AP 786-081 786-003 786-10T
82022-458EA 440.37 USD InStock
CA82022-458 82022-458 CA10118-294 CA89167-702 82022-460 10063-114 89167-702 10063-112 CA10118-296 CA95043-412 95043-412 CA95043-410 95043-410 CA82022-460
Femtogram Sensitive Chemiluminescence Detection, G-Biosciences
Assays ELISAs
The femtoLUCENT™ PLUS Reagent Kit facilitates the quick and sensitive non-isotopic detection of proteins on transfer membranes.
  • Intense light emission with low background: high signal ratio
  • Low femtogram detection (10 to 15), allows detection of >10 fg protein on a dot blot and >1 pg on a Western blot
  • Supplied with a novel blocking agent which allows a rapid blocking step and produces a clear background
  • Suitable for nitrocellulose and PVDF membranes
A femtogram level sensitive immunodetection system allows users the option to detect even hard-to-detect low abundance proteins. This immunodetection system is based on an innovative formulation of luminol and 1,2 dioxetane supersensitive detection reagent and a unique, proprietary combination of blocking agent (NAP-BLOCKER™) and washing buffers to ensure low background and high signal to background ratio. Optional horseradish peroxidase (HRP) or alkaline phosphatase (AP) conjugated secondary antibodies against mouse, rabbit, human, rat and goat are also offered for a complete chemiluminescence detection system.
femtoLUCENT™ PLUS is offered as a complete immunodetection system with all of the critical reagents needed for Western blots or dot-blot analysis or as a reagent kit for those who prefer to customise their detection methods with the choice of their own secondary antibody conjugates.
Ordering information: femtoLUCENT™ PLUS reagent kit includes detection reagents for HRP or AP, NAP-BLOCKER™, a non animal protein blocking agent, femto-TBST™ washing buffer, and optional secondary antibodies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty