Would you like to log in again?
SNAPTOP VIAL & CAP EPP AMB
Supplier: MicroSolv Technology Corp
BASIK* 1.5 ml Snaptop vial and cap eppendorf amber^ Snap/crimp top autosampler vials, 12x32mm and blue AQR snap caps with locked in fitted ultra high purity silicone/PTFE septa. 100/pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SNAPTOP VIAL & CAP EPP AMB 10063-376 9502C-T-1AP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty