Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide solution 15%

Supplier: HARDY DIAGNOSTICS

Z262
10063-756EA 90.46 USD InStock
10063-756
Hydrogen peroxide solution 15%
Hydrogen peroxide
A high strength 15% solution of hydrogen peroxide especially recommended for the detection of catalase activity in anaerobic bacteria.


Formula: H₂O₂
MW: 34.01 g/mol
MDL Number: MFCD00011333
CAS Number: 7722-84-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty