Would you like to log in again?
PermaBlue Plus/AP, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K058 K058 K058-110
10064-752EA 156.8 USD InStock
10064-752 10064-754 CA10064-752
PermaBlue Plus/AP, Diagnostic BioSystems
Stains Electrophoresis Stains
PermaBlue Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase
It has increase stability of the working solution and produces a distinct bright blue color. It is insoluble in alcohol and xylene substitutes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty