Would you like to log in again?
Buffers (10X) For Heat Induced Epitope Recovery, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K043 K 044 K038 K038
10064-760EA 470.69 USD InStock
10064-760 10064-758 CA99990-102 10064-756
Buffers (10X) For Heat Induced Epitope Recovery, Diagnostic BioSystems
Buffers Antibody Purification Buffers
The is a heavy metal free antigen unmasking solution provided in a convenient 10X concentrate
Use of this antigen recovery buffer avoids unnecessary heavy metal exposure to lab personal and handling and disposal issues.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty