Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pro1020 Field DO, pH/ORP and Temp Handheld Meter, YSI

Supplier: YSI

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

6051020
10064-896EA 1355.03 USD InStock
10064-896
Pro1020 Field DO, pH/ORP and Temp Handheld Meter, YSI
Electrochemical Meters pH and Multi-Parameter Meters
Rugged and reliable, the YSI Pro1020 provides everything you need in a handheld instrument for the measurement of either pH or ORP along with temperature, and dissolved oxygen.

User-replaceable sensors and cables along with convenient calibration routine feature makes it extremely user friendly. Mil-spec (military spec) connectors, IP67 waterproof and rubber over molded case, and 1-meter drop tests ensure the instrument remains in your hands to provide years of sampling even in the harshest field conditions. The feature set of the Pro1020 results in a low total cost of ownership over the life of the product and proves that it is smarter, tougher, and more versatile.

The Pro1020 simultaneously measures dissolved oxygen, pH/ORP, and temperature with the following features: User-replaceable cables and sensors provide versatility, reduce down time and reduce overall cost of ownership. Available with 1- 4- 10- 20- and 30-m cable lengths standard (cable management kit included on all cables 4-m and longer). Smart calibration; the instrument remembers the previous calibration values and simply walks you through the routine. Stores 50 data sets; no need to write down data. Large graphic, backlit display and glow in the dark keypad. Usable in any lighting condition. The display is also visible when wearing polarized sunglasses. Tough. IP67, impact-resistant, waterproof case. Waterproof without the battery cover as well. Rubber over molded case provides extra durability. Military spec connectors are unsurpassed for ruggedness. Multiple languages include English, Spanish, French, and Portuguese. Choose either polarographic or galvanic DO sensors. Quick response times; 95% DO response time in approximately 8 seconds with standard membrane (fastest response time in the market). 3-year instrument; 2-year cable; 1-year sensor warranty (Galvanic DO is 6-months).

Certifications: RoHS. CE. WEEE. 1-meter drop test. IP67 (even with the battery cover off).

Ordering information: When ordering the Pro1020 you must order the instrument, cable, and dissolved oxygen (Polarographic of Galvanic), pH or ORP sensor separately, allowing you to choose the system that best fits your application. Temperature is part of the cable you order and included in the cable purchase. Contact your VWR representative for more information. Warranty includes 3-year instrument; 2-year cable; 1-year pH and ORP sensors (Galvanic DO is 6-months).

Delivery information: Cable length: 1- 4- 10- 20- and 30-m lengths (3.28, 13.1, 32.8, 65.6, and 98.4 ft.)

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty