Would you like to log in again?
Urea Agar Slant, Hardy Diagnostics
Supplier: Hardy Diagnostics
R42 L65 R42
10753-980PK 86.82 USD InStock
CA101380-976 10753-980 10065-304
Urea Agar Slant, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Urea Agar Slant (Christensen’s Urea) is useful for the detection of urease production in bacteria.
  • Detects urease production in bacteria
  • Rapid growth
  • Fast reaction time
The slant contains urea and phenol red as the pH indicator
Proteus species rapidly hydrolyze urea, and a positive reaction is usually seen within one to six hours. Other organisms perform this reaction slower and may require 24 to 48 hours or longer incubation time.
The pH indicator turns from pale yellow to pink-red when cabable organisms hydrolyze urea form ammonia and turn the medium alkaline. The reduced buffer content and peptone in this medium promote more rapid growth and reaction time for many members of the Enterobacteriaceae. Dextrose is included in the formulation to stimulate urease activity in organisms that hydrolyze urea slowly, and to exclude false-negative reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty